missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mutexil™ Powder-Free Natural Vinyl Gloves
Mutexil™ Powder-Free Natural Vinyl Gloves are disposable and come in a range of sizes. Tested for food contact.
Marke: Mutexil™ 6663190XL
Additional Details : Gewicht : 0.01000kg
Beschreibung
- Powder-free
- Natural vinyl
- Supplied in convenient dispenser boxes
- Non-sterile
Spezifikation
Natural | |
Vinyl | |
Powder-free | |
AQL 1.5, EN 455-1-2&3, EN420, CE | |
Ambidextrous |
Beaded | |
X-Large | |
Non-sterile | |
Yes | |
Yes |