missing translation for 'onlineSavingsMsg'
Learn More

Kalium 3,5-Dinitro-2 -Methylphenyltrifluoroborat, 96 %, Thermo Scientific™

Catalog Number 15460536
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
850623-72-2
C7H5BF3KN2O4
288.031
MFCD04115767
WATHPRZGMORMCV-UHFFFAOYSA-N
potassium 3,5-dinitro-2-methylphenyl trifluoroborate, potassium trifluoro 2-methyl-3,5-dinitrophenyl borate, potassium trifluoro 2-methyl-3,5-dinitrophenyl boranuide, potassium 3,5-dinitro-2-methylphenyltrifluoroborate, c7h5bf3n2o4.k, potassium trifluoro 2-methyl-3,5-dinitrophenyl borate 1-
44886960
Kalium;Trifluoro-(2-Methyl-3,5-dinitrophenyl)Boranuid
[B-](C1=CC(=CC(=C1C)[N+](=O)[O-])[N+](=O)[O-])(F)(F)F.[K+]
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
850623-72-2
C7H5BF3KN2O4
288.031
MFCD04115767
WATHPRZGMORMCV-UHFFFAOYSA-N
potassium 3,5-dinitro-2-methylphenyl trifluoroborate, potassium trifluoro 2-methyl-3,5-dinitrophenyl borate, potassium trifluoro 2-methyl-3,5-dinitrophenyl boranuide, potassium 3,5-dinitro-2-methylphenyltrifluoroborate, c7h5bf3n2o4.k, potassium trifluoro 2-methyl-3,5-dinitrophenyl borate 1-
44886960
Kalium;Trifluoro-(2-Methyl-3,5-dinitrophenyl)Boranuid
[B-](C1=CC(=CC(=C1C)[N+](=O)[O-])[N+](=O)[O-])(F)(F)F.[K+]

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material Potassium 3,5-dinitro-2-methylphenyltrifluoroborate
Schmelzpunkt 206°C to 210°C
Menge 250 mg
Formelmasse 288.03
Reinheit (%) 96%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'