missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novex™ Positope™ Control Protein
Beschreibung
The Positope™ Control Protein is designed to be used as a positive control in western blotting for a variety of antibodies. The control protein is a 53 kDa highly purified (>95%) recombinant protein that contains seven epitope tags. Validated applications for the Positope Control Protein are Western Blot against the primary antibodies listed in the manual. By using Positope™ Protein as a positive control, you can assess the performance of your western blots, no matter which of the epitopes you are using.
Control Protein Attributes:
Applications: Validated applications for Positope™ Control Protein are Western blotting using the primary antibodies mentioned in the manual. Recommended loading amount for Western blotting: 100 ng per lane.
Protein Subtype: Tagged Protein Control
Product Size: Positope™ Control Protein is supplied as a 5μg pack size.
Spezifikation
Spezifikation
| Zur Verwendung mit (Anwendung) | Assay Standard |
| Menge | 5 μg |
| Inhalt und Lagerung | Each vial contains 5 μg of Positope™ Control Protein at a concentration of 25 ng/μl in reducing SDS-PAGE sample buffer. Store the protein at +4°C. For long-term storage, store at -20°C. The protein is guaranteed stable for 6 months when stored properly. |
| Proteinfamilie | Other Proteins |
| Proteinform | Recombinant |
| Protein-Subtyp | Tagged Protein Controls |
| Produktlinie | Positope |
For Research Use Only. Not for use in diagnostic procedures.
Name des Produkts
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.
Haben Sie Verbesserungsvorschläge?