missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific™ POROS™ CaptureSelect™ AAV8 Affinity Resin Artikelnummer.: 18446678
Verfügbar ab: 04-08-2025
Nur noch null auf Lager
Zum Warenkorb hinzufügen

Thermo Scientific™ POROS™ CaptureSelect™ AAV8 Affinity Resin

Artikelnummer. 18446678 Alle ansehen Thermo Scientific Produkte
25 mL, 25 Milliliter
Click to view available options
Menge:
10 mL
25 mL
50 mL
250 mL
Packungsgröße:
10 Milliliter
25 Milliliter
250 Milliliter
50 Milliliter
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Artikelnummer. 18446678

Marke: Thermo Scientific™ A30790

Nur noch null auf Lager
Verfügbar ab: 04-08-2025
Zum Warenkorb hinzufügen
Nur noch null auf Lager
Zum Warenkorb hinzufügen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Thermo Scientific™ POROS™ CaptureSelect™ AAV8 high performance affinity resin is engineered to address the high selectivity and capacity requirements needed for the purification of viral vectors used for gene therapy applications and is specifically intended for the purification of adeno-associated virus type 8 (AAV8).

POROS CaptureSelect AAV8 high performance affinity resin is engineered to address the high selectivity and capacity requirements needed for the purification of viral vectors used for gene therapy applications and is specifically intended for the purification of adeno-associated virus type 8 (AAV8).

Features of POROS CaptureSelect AAV8 resin include:

  • One-step AAV8 purification from crude material with high purity and yield
  • High specificity and capacity, which helps reduce the process volume significantly for subsequent steps and maximizes yield
  • Low operating back pressure and linear pressure versus flow responses driving flexible scalability
  • Superior capacity maintained at high flow rates
  • Rigid polymeric bead with covalent surface chemistry for easier handling and packing, with superior physical and chemical stability enabling a more robust downstream purification process
  • Non-animal derived—the ligand is a 13-kDa single-domain fragment that comprises the 3 CDRs that form the antigen-binding domain and is efficiently produced by the yeast Saccharomyces cerevisiae in a production process free of animal components.

POROS CaptureSelect AAV8 affinity resin specifically binds AAV8 and enables a robust, fast, and efficient purification process with excellent purity and yield obtained in one step. The use of this affinity resin will enable significant improvement to downstream processing by reducing the purification steps and maximizing productivity, for scalability and processing consistency.

Main characteristics:

  • Matrix: cross-linked poly(styrene-divinylbenzene)
  • Average particle size: 50 μm
  • Ligand: POROS CaptureSelect AAV8 affinity ligand
  • Ligand coupling method: carbonyldiimidazole (CDI)
  • Binding capacity: >1 x 1013 genome copies/mL of resin
  • Elution conditions: acidic elution with 0.1 M glycine, pH 3.0
  • Mechanical resistance: 100 bar (1,450 psi; 10 MPa)
  • Formulation buffer: 18% (v/v) ethanol

Spezifikation

Bindungsvermögen >1 x 1013 genome copies/mL of resin
Matrix Cross-linked poly(styrene-divinylbenzene)
Inhalt und Lagerung Store at 2°C to 8°C.
Versandbedingung Nasseis
Partikelgröße 50 μm
Form Flüssige Suspension
Produktlinie CaptureSelect™, POROS™
Menge 25 mL
Typ Harz-
Ligandentyp AAV8 affinity ligand

Zertifikate

Eine Chargennummer ist erforderlich, um Ergebnisse für Zertifikate anzuzeigen. Um Ihre Chargennummer zu früheren Bestellungen zu finden, können Sie unsere Auftragsverfolgung nutzen.

5 Ergebnisse gefunden
Chargennummer Art des Zertifikats Datum Artikelnummer
ChargennummerAAV8-029 Art des ZertifikatsAnalysenzertifikat Datum25/06/2024 ArtikelnummerA30790
ChargennummerAAV8-028 Art des ZertifikatsAnalysenzertifikat Datum06/12/2023 ArtikelnummerA30790
ChargennummerAAV8-022 Art des ZertifikatsAnalysenzertifikat Datum01/04/2021 ArtikelnummerA30790
ChargennummerAAV8-020 Art des ZertifikatsAnalysenzertifikat Datum08/01/2021 ArtikelnummerA30790
ChargennummerAAV8-019 Art des ZertifikatsAnalysenzertifikat Datum26/07/2019 ArtikelnummerA30790
Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts
Thermo Scientific™ POROS™ CaptureSelect™ AAV8 Affinity Resin >

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt