Learn More
Tocris Bioscience™ Pomalidomide 4'-PEG1-amine
Click to view available options
Menge:
25 mg
Packungsgröße:
25 Milligramm
Beschreibung
Pomalidomide 4'-PEG1-amine is a functionalized cereblon ligand for PROTAC™ research and development; incorporates an E3 ligase ligand plus a PEG1 linker with terminal amine ready for conjugation to a target protein ligand. Part of a range of functionalized tool molecules for PROTAC R&D. PROTAC™ is a registered trademark of Arvinas Operations, Inc., and is used under license.
Spezifikation
Spezifikation
| Chemischer Name oder Material | 4-[[2-(2-Aminoethoxy)ethyl]amino]-2-(2,6-dioxo-3-piperidinyl)-1 H-isoindole-1,3(2 H)-dione hydrochloride |
| CAS | 2154342-56-8 |
| Menge | 25 mg |
| Ziel | Degrader Building Blocks |
| Summenformel | C17H20N4O5.HCl |
| Reinheit | 0.95 |