missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polysulfid-Ölstandard: 40.000 μg/g S in Mineralöl, VHG™
Description
Polysulfid-Ölstandard: 40.000 μg/g S in Mineralöl
Specifications
Specifications
| Chemischer Name oder Material | Sulfur |
| CAS | 7704-34-9 |
| Bezeichnung der Analyten oder Bestandteile | Sulfur |
| Versandbedingung | Raumtemperatur |
| Summenformel | S |
| Konzentration | 40.000 μg/g |
| InChI-Formel | InChI=1S/S |
| SMILES | [S] |
| IUPAC-Name | Schwefel |
| Solvent or Matrix | 13 cSt Mineralöl |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?