missing translation for 'onlineSavingsMsg'
Learn More

Podocalyxin Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

133.80 CHF - 381.34 CHF

Spezifikation

Antigen Podocalyxin
Konzentration 0.26 mg/mL
Anwendungen Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin)
Klassifikation Polyclonal
Konjugat Unconjugated
Mehr Spezifikationen ansehen

Produkte 2
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
16838204
Unterlagen ansehen Aktionsdetails
Proteintech
18150-1-AP-20UL
20 μL
133.80 CHF
20 Mikroliter
Alternative Produkte anzeigen
Sparen Sie bis zu 
»
Verfügbar ab: 17-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
 
16828204
Unterlagen ansehen Aktionsdetails
Proteintech
18150-1-AP-150UL
150 μL
381.34 CHF
150 Mikroliter
Alternative Produkte anzeigen
Sparen Sie bis zu 
»
Verfügbar ab: 17-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
 
Beschreibung

Beschreibung

PODXL encodes a member of the sialomucin protein family. The encoded protein was originally identified as an important component of glomerular podocytes. Podocytes are highly differentiated epithelial cells with interdigitating foot processes covering the outer aspect of the glomerular basement membrane. Other biological activities of the encoded protein include: binding in a membrane protein complex with Na+/H+ exchanger regulatory factor to intracellular cytoskeletal elements, playing a role in hematopoetic cell differentiation, and being expressed in vascular endothelium cells and binding to L-selectin.
Spezifikation

Spezifikation

Podocalyxin
Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Mouse, Human
O00592, Q9R0M4
27205, 5420
Podocalyxin Fusion Protein Ag12844
Primary
-20°C
PODXL
0.26 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
GCTM 2 antigen, Gp200, PC, PCLP, PCLP 1, PCLP1, Podocalyxin, podocalyxin like, Podocalyxin like protein 1, PODXL, RCC marker, Renal Cell Carcinoma marker, TRA 1 81,TRA 1 60, TRA-1-60
Podxl
IgG
Antigen Affinity Chromatography
Antibody
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Product Certifications