Learn More
PNP Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA551616
Additional Details : Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: YTYEDYKNTA EWLLSHTKHR PQVAIICGSG LGGLTDKLTQ AQIFDYGEIP NFPRSTVPGH AGRLVFGFLN GRACVMMQGR FHMYEGYPLW KVTFPVRVFH LLGVDTLVVT NAAGGLNPKF EVGDIMLIRD HINLPGFSGQ NPLRGPNDER GAPHR Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 87%.
PNP Polyclonal antibody specifically detects PNP in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
PNP | |
Polyclonal | |
Unconjugated | |
PNP | |
AL024301; AU015798; endothelial dysfunction inducing protein; epididymis secretory sperm binding protein Li 156an; HEL-S-156an; inosine phosphorylase; inosine phosphorylase {ECO:0000250; Inosine-guanosine phosphorylase; Np; Np-1; Np-2; nucleoside phosphorylase; PNP; PNP {ECO:0000250; Pnp1; PRO1837; PUNP; purine nucleoside phosphorylase; purine nucleoside phosphorylase {ECO:0000250; purine-nucleoside phosphorylase; purine-nucleoside phosphorylase 1; purine-nucleoside:orthophosphate ribosyltransferase; UniProtKB:P00491} | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
18950, 290029, 4860 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P00491, P23492, P85973 | |
PNP | |
Recombinant protein corresponding to Human PNP | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Research Use Only