missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ PMF1 Recombinant Protein
Marke: Abnova™ H00011243-P01.10ug
Additional Details : Gewicht : 0.00010kg
Beschreibung
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Spezifikation
AAH33656 | |
Solution | |
11243 | |
PMF1 (Human) Recombinant Protein (P01) | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Wheat Germ (in vitro) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PMF1 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
45.3 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
MAEASSANLGSGCEEKRHEGSSSESVPPGTTISRVKLLDTMVDTFLQKLVAAGSYQRFTDCYKCFYQLQPAMTQRIYDKFIAQLQTSIREEISDIKEEGNLEAVLNALDKIVEEGKVRKEPAWCEAQRCRVQQRCSLCVQAGGQRGSEETQALPVSMAGSPSPLPGSPGAQEGGV | |
PMF1 | |
Human | |
Yes |