missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLEK Mouse anti-Human, Porcine, Clone: 1D5C2, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech 66431-1-IG-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
Major protein kinase C substrate of platelets.Spezifikation
PLEK | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P08567 | |
PLEK | |
PLEK Fusion Protein Ag3187 | |
20 μL | |
Primary | |
Human, Porcine | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1D5C2 | |
Unconjugated | |
PLEK | |
P47, Platelet 47 kDa protein, pleckstrin, PLEK | |
Mouse | |
Protein G | |
RUO | |
100516257, 5341 | |
Antibody | |
IgG1 |