Learn More
Thermo Scientific™ Human PKMYT1, GST Tag Recombinant Protein
This PKMYT1 recombinant human protein (full length) was expressed in insect cells. PKMYT1 serine/threonine kinase is a member of a family of neural specific, zinc finger-containing DNA-binding proteins which binds to the promoter regions of proteolipid proteins of the central nervous system.
Marke: Thermo Scientific™ A33387
Additional Details : Gewicht : 0.10000kg
Beschreibung
- This PKMYT1 recombinant human protein (full length) was expressed in insect cells
- PKMYT1 serine/threonine kinase is a member of a family of neural specific, zinc finger-containing DNA-binding proteins
- MYT1 of the oligodendrocyte lineage, along with a closely related CCHC zinc finger, is expressed in developing neurons in the mammalian central nervous system
- PKMYT1 recombinant human protein is also available in our SelectScreen™ Kinase Profiling Service
- Validated for use with the LanthaScreen™ Eu Kinase Binding assay
- Available in multiple pack sizes for convenient use
- Also available in our SelectScreen Kinase Profiling Service
Spezifikation
NP_004194 | |
80.8 kDa | |
WEE Kinase Family | |
Human | |
GST Tag | |
Dry Ice | |
Kinases & Inhibitors | |
Liquid |
Kinase Assay | |
1 mg | |
Full Length | |
Recombinant | |
Storage Buffer: 50 mM Tris-HCl, pH 7.5, 50 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol | |
Baculovirus | |
Recombinant, Full Length |