missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PKC delta Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 19132-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
Protein kinase C (PKC) is a family of serine- and threonine-specific protein kinases that can be activated by calcium and the second messenger diacylglycerol. PKC family members phosphorylate a wide variety of protein targets and are known to be involved in diverse cellular signaling pathways. PKC family members also serve as major receptors for phorbol esters, a class of tumor promoters. Each member of the PKC family has a specific expression profile and is believed to play distinct roles in cells. The protein encoded by this gene is one of the PKC family members. Studies both in human and mice demonstrate that this kinase is involved in B cell signaling and in the regulation of growth, apoptosis, and differentiation of a variety of cell types. Alternatively spliced transcript variants encoding the same protein have been observed.Spezifikation
PKC delta | |
Polyclonal | |
Unconjugated | |
PRKCD | |
MAY1, PKC delta, PKCD, PRKCD, Protein kinase C delta type, protein kinase C, delta | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
170538, 18753, 5580 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P09215, P28867, Q05655 | |
Prkcd | |
PKC delta Fusion Protein Ag5631 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |