missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PKC delta Monoclonal Antibody (8E12), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA519701
Additional Details : Gewicht : 0.00010kg
Beschreibung
Peptide Sequence: DILEKLFERE PTKRLGVTGN IKIHPFFKTI NWTLLEKRRL EPPFRPKVKS PRDYSNFDQE FLNEKARLSY SDKNLIDSMD QSAFAGFSFV NPKFEHLLED
PKC delta Monoclonal antibody specifically detects PKC delta in Human samples. It is validated for ELISA, Immunocytochemistry, Western Blot.Spezifikation
PKC delta | |
Monoclonal | |
1.0 mg/mL | |
PBS with no preservative; pH 7.4 | |
Q05655 | |
PRKCD | |
PRKCD (NP_006245, 577 a.a. ∽ 676 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
ELISA, Immunocytochemistry, Western Blot | |
8E12 | |
Unconjugated | |
PRKCD | |
1-May; AI385711; ALPS3; CVID9; D14Ertd420e; kinase nPKC-delta; KPCD; MAY1; nPKC-delta; PKC[d]; Pkcd; PKCdelta; Prkcd; protein kinase C delta; protein kinase C delta type; Protein kinase C delta type catalytic subunit; Protein kinase C delta type regulatory subunit; protein kinase C delta variant IX; protein kinase C delta VIII; protein kinase C, delta; protein kinase C, delta IV; protein kinase C, delta V; protein kinase C[d]; SDK1; Sphingosine-dependent protein kinase-1; Tyrosine-protein kinase PRKCD | |
Mouse | |
Protein A | |
RUO | |
5580 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |