Learn More
PKC alpha Mouse anti-Human, Mouse, Rat, Clone: 1D11F4, Proteintech
Mouse Monoclonal Antibody
Marke:  Proteintech 66421-1-IG-20UL
 Lernen Sie weitere Varianten dieses
Artikelnummer. 16838285
  • 136.48 CHF / 20 Mikroliter
Nur noch null auf Lager
Lager vorrätig
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
Protein kinase C (PKC) is a family of serine- and threonine-specific protein kinases that can be activated by calcium and the second messenger diacylglycerol. PKC family members phosphorylate a wide variety of protein targets and are known to be involved in diverse cellular signaling pathways. PKC family members also serve as major receptors for phorbol esters, a class of tumor promoters. Each member of the PKC family has a specific expression profile and is believed to play a distinct role in cells. The protein encoded by this gene is one of the PKC family members. This kinase has been reported to play roles in many different cellular processes, such as cell adhesion, cell transformation, cell cycle checkpoint, and cell volume control. Knockout studies in mice suggest that this kinase may be a fundamental regulator of cardiac contractility and Ca(2+) handling in myocytes.
Spezifikation
Mehr anzeigen
Produktvorschläge
Produktvorschläge
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate
Aktionen
Aktionen
Berichtigung von Produktinhalten
Name des Produkts
PKC alpha Mouse anti-Human, Mouse, Rat, Clone: 1D11F4, Proteintech > 20 μL; Unconjugated
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado