missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIP5K1B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
136.48 CHF - 385.92 CHF
Spezifikation
| Antigen | PIP5K1B |
|---|---|
| Konzentration | 0.34 mg/mL |
| Anwendungen | Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation |
| Klassifikation | Polyclonal |
| Konjugat | Unconjugated |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|
16870294
|
Proteintech
12541-1-AP-20UL |
20 μL |
136.48 CHF
20 Mikroliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
|
16860294
|
Proteintech
12541-1-AP-150UL |
150 μL |
385.92 CHF
150 Mikroliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
Protein kinases are enzymes that transfer a phosphate group from a phosphate donor, generally the g phosphate of ATP, onto an acceptor amino acid in a substrate protein. By this basic mechanism, protein kinases mediate most of the signal transduction in eukaryotic cells, regulating cellular metabolism, transcription, cell cycle progression, cytoskeletal rearrangement and cell movement, apoptosis, and differentiation. With more than 500 gene products, the protein kinase family is one of the largest families of proteins in eukaryotes. The family has been classified in 8 major groups based on sequence comparison of their tyrosine (PTK) or serine/threonine (STK) kinase catalytic domains.Spezifikation
| PIP5K1B | |
| Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Rat, Human, Mouse | |
| O14986, P70181, Q5CZZ9 | |
| 18719, 309419, 8395 | |
| PIP5K1B Fusion Protein Ag3261 | |
| Primary | |
| -20°C | |
| PIP5K1B |
| 0.34 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| MSS4, PIP5K1 beta, PIP5K1B, PIP5KIbeta, Protein STM 7, PtdIns(4)P 5 kinase 1 beta, STM7 | |
| Pip5k1b | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts