missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PINX1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 12368-1-AP-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
Valosin containing protein (VCP), also designated TERA (for transitional endoplasmic reticulum ATPase), is a member of the AAA family of ATPases, which are involved in a variety of cellular activities. VCP is involved in a variety of membrane functions and in the regulation of the cell cycle. VCP associates with ubiquitinated IkB-a as well as with the 26S proteosome, indicating a potential role for VCP in the proteosome-mediated degradation of IkB-a. NVL (nuclear valosin-containing protein-like), also known as NVLp, is an 856 amino acid nuclear protein belonging to the AAA ATPase family. Implicated in ATP-dependent nuclear processes and ribosome synthesis, NVL exists as three alternatively spliced isoforms designated NVL isoform 1 (NVLp.2), NVL isoform 2 (NVLp.1) and NVL isoform 3. Widely expressed, NVL is found at highest levels in pancreas, retina, heart, skeletal muscle and placenta.Spezifikation
PINX1 | |
Polyclonal | |
Unconjugated | |
PINX1 | |
LPTL, LPTS, PIN2 interacting protein 1, Pin2 interacting protein X1, PINX1, Protein 67 11 3, TRF1 interacting protein 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
305963, 54984, 72400 | |
Antibody | |
IgG |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
A4L691, Q96BK5, Q9CZX5 | |
PINX1 | |
PINX1 Fusion Protein Ag3043 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Liquid |