Learn More
Pilocarpinhydrochlorid, 99,3%, MP Biomedicals™
Artikelnummer. 11427122 Alle ansehen MP Biomedicals Produkte
Change view
Click to view available options
Menge:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge
11427122 1 g
11437122 5 g
11447122 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 11427122 Lieferant MP Biomedicals Lieferanten-Nr. 0215189201
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Chemische Identifikatoren
CAS 54-71-7
Summenformel C11H17ClN2O2
Molekulargewicht (g/mol) 244.72
MDL-Nummer MFCD00012722
InChI-Schlüssel RNAICSBVACLLGM-GNAZCLTHSA-N
Synonym pilocarpine hydrochloride, +-pilocarpine hydrochloride, pilocarpal, pilocar, pilocar smp, almocarpine, pilomiotin, pilovisc, pilocel, pilocarpine hcl
PubChem CID 5909
IUPAC-Name (3S,4R)-3-Ethyl-4-[(1-methyl-1H-imidazol-5-yl)methyl]oxolan-2-on-chlorid
SMILES [H+].[Cl-].CC[C@H]1[C@@H](CC2=CN=CN2C)COC1=O
Spezifikation
Chemischer Name oder Material Pilocarpine hydrochloride
Prozentgehaltsbereich 0.993
Menge 1 g
Formelmasse 244.7
Physikalische Form Crystalline