missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pikachurin Mouse anti-Human, Clone: 1C6D12, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech 66232-1-IG-20UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
This monoclonal antibody detects two bands of 111 kDa and 55-60 kDa. The lower-molecular-weight band has been reported and likely represents a fragment resulting from cleavage of Pikachurin.
Involved in both the retinal photoreceptor ribbon synapse formation and physiological functions of visual perception. Necessary for proper bipolar dendritic tip apposition to the photoreceptor ribbon synapse. Promotes matrix assembly and cell adhesiveness.Spezifikation
| Pikachurin | |
| Monoclonal | |
| 2 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q63HQ2 | |
| EGFLAM | |
| Pikachurin Fusion Protein Ag6362 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| 1C6D12 | |
| Unconjugated | |
| EGFLAM | |
| Agrin like protein, AGRINL, AGRNL, EGFLAM, Pikachurin | |
| Mouse | |
| Protein G | |
| RUO | |
| 133584 | |
| -20°C | |
| Liquid |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion