Learn More
Phosphorsäuretris(4-methylphenyl)ester, TRC
Specifications
Specifications
| Chemischer Name oder Material | Tri-p-cresyl phosphate |
| CAS | 78-32-0 |
| Summenformel | C21 H21 O4 P |
| Menge | 1 g |
| Synonym | Phosphoric acid','tris(4-methylphenyl) ester (9CI','ACI), Phosphoric acid','tri-p-tolyl ester (8CI), NSC 2181, p-Tolyl phosphate (C7H7O)3PO, Tri-p-cresyl phosphate, Tri-p-tolyl phosphate, Tris(4-methylphenyl) phosphate, Tris(p-cresyl) phosphate, Tris(p-methylphenyl) phosphate, Tri-4-cresyl phosphate |
| InChI-Formel | InChI=1S/C21H21O4P/c1-16-4-10-19(11-5-16)23-26(22,24-20-12-6-17(2)7-13-20)25-21-14-8-18(3)9-15-21/h4-15H,1-3H3 |
| SMILES | Cc1ccc(OP(=O)(Oc2ccc(C)cc2)Oc3ccc(C)cc3)cc1 |
| IUPAC-Name | Tris(4-Methylphenyl)phosphat |
| Molekulargewicht (g/mol) | 368.36 |
| Formelmasse | 368.1177 |