missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-PLK1 (Ser326) Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 19553-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
The antibody is specific to Ser(326)-phospho.
PLK1 (Polo Like Kinase 1) is a Protein Coding gene. Diseases associated with PLK1 include Transient Tic Disorder and Monoclonal Paraproteinemia. Among its related pathways are Integrated Breast Cancer Pathway and Cell Cycle, Mitotic. Gene Ontology (GO) annotations related to this gene include transferase activity, transferring phosphorus-containing groups and protein tyrosine kinase activity. An important paralog of this gene is PLK3. This gene encodes a protein which contains a conserved nuclear localization signal.Spezifikation
Phospho-PLK1 (Ser326) | |
Polyclonal | |
Unconjugated | |
PLK1 | |
PLK, PLK 1, PLK1, Polo like kinase 1, STPK13 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
18817, 25515, 5347 | |
Antibody | |
IgG |
Western Blot, Immunohistochemistry (Paraffin) | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P53350, Q07832, Q62673 | |
Plk1 | |
Peptide | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Liquid |