missing translation for 'onlineSavingsMsg'
Learn More

Phospho-PLCG2 (Tyr759) Mouse anti-Human, Mouse, APC, Clone: 4NPRN4, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody

Marke:  Invitrogen 17-9866-41

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15983224

  • 133.00 CHF / 25 Stück


Verfügbar ab
30-09-2020
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Phospho-PLCG2 (Tyr759) Monoclonal antibody specifically detects Phospho-PLCG2 (Tyr759) in Human, Mouse samples. It is validated for Flow Cytometry
Spezifikation

Spezifikation

Phospho-PLCG2 (Tyr759)
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P16885, Q8CIH5
1-phosphatidylinositol 4,5-bisphosphate phosphodiesterase gamma-2; APLAID; FCAS3; Phosphoinositide phospholipase C-gamma-2; Phospholipase C-gamma-2; Phospholipase C-IV; PLC gamma 2; PLC gamma2; PLC- gamma2; PLC gamma-2; Plcg2; Plcg-2; PLC-gamma; PLCgamma2; PLC-gamma-2; PLC-IV
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human, Mouse
Flow Cytometry
4NPRN4
APC
Plcg2
Liquid
Plcg2
IgG1, kappa
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
234779, 5336
Videos
Dokumentation