missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-PKC delta (Ser664) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen 44976G
Additional Details : Gewicht : 0.50000kg
Beschreibung
Phospho-PKC delta (Ser664) Polyclonal antibody specifically detects Phospho-PKC delta (Ser664) in Human, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
Phospho-PKC delta (Ser664) | |
Polyclonal | |
Dulbecco's PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.3 | |
P09215, Q05655 | |
PRKCD | |
The antiserum was produced against a chemically synthesized phosphopeptide derived from a region of human PKCd that contains serine 664. | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
PRKCD | |
1-May; AI385711; ALPS3; CVID9; D14Ertd420e; kinase nPKC-delta; KPCD; MAY1; nPKC-delta; PKC[d]; Pkcd; PKCdelta; Prkcd; protein kinase C delta; protein kinase C delta type; Protein kinase C delta type catalytic subunit; Protein kinase C delta type regulatory subunit; protein kinase C delta variant IX; protein kinase C delta VIII; protein kinase C, delta; protein kinase C, delta IV; protein kinase C, delta V; protein kinase C[d]; SDK1; Sphingosine-dependent protein kinase-1; Tyrosine-protein kinase PRKCD | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
170538, 5580 | |
-20°C | |
Liquid |