missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phospho-MUSK (Tyr755) Polyclonal Antibody
GREENER_CHOICE

Artikelnummer. 16671825
Change view
Click to view available options
Menge:
100 μL
Packungsgröße:
100 Mikroliter
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
16671825 100 μL 100 Mikroliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 16671825 Lieferant Invitrogen™ Lieferanten-Nr. PA5105382

om dit product te kopen Registreer vandaag om een webaccount aan te maken

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of MuSK only when phosphorylated at Tyr755.

MUSK plays an important and selective role in many aspects of skeletal muscle development and function through the initial step of postsynaptic differentiation and formation of end-plate bands in cooperation with Rapsyn. Communication between cells is often mediated by receptors on the surface of the target cell that are triggered by specific ligands released by other cells. Receptor tyrosine kinase activity is strictly regulated by the binding of the appropriate ligand to the extracellular active site of the receptor. MuSK has been shown to be part of the agrin signaling complex and plays an important role in the development and architectural maintenance of the neuromuscular junction.
TRUSTED_SUSTAINABILITY

Spezifikation

Antigen Phospho-MUSK (Tyr755)
Anwendungen Immunohistochemistry (Paraffin), Western Blot
Klassifikation Polyclonal
Konzentration 1 mg/mL
Konjugat Unconjugated
Zusammensetzung PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gen MUSK
Gen-Zugriffsnummer O15146, Q61006, Q62838
Gen-Alias CMS9; FADS; Mdk4; MGC126323; MGC126324; Mlk; muscle associated receptor tyrosine kinase; muscle localized kinase 2; muscle specific kinase (neural fold/somite kinase 1); muscle, skeletal receptor tyrosine protein kinase; Muscle, skeletal receptor tyrosine-protein kinase; muscle, skeletal, receptor tyrosine kinase; muscle-specific kinase receptor; muscle-specific protein kinase secretory isoform; muscle-specific tyrosine kinase receptor MuSK; muscle-specific tyrosine protein kinase receptor; Muscle-specific tyrosine-protein kinase receptor; MuSK; Nsk1; Nsk2; Nsk3; RP23-170A3.1
Gensymbole MUSK
Wirtsspezies Rabbit
Immunogen A synthesized peptide derived from human MUSK(Accession O15146), corresponding to amino acid residues around phosphorylated Tyr755.
Reinigungsverfahren Sequential chromatography
Menge 100 μL
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 18198, 4593, 81725
Zielspezies Human, Mouse, Rat
Inhalt und Lagerung -20°C
Produkttyp Antibody
Form Liquid
Isotype IgG
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.