missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-JAK2/JAK3 (Tyr966, Tyr939) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA567508
Additional Details : Gewicht : 0.01000kg
Beschreibung
Predicted to react with Rat samples.
Janus Activating Kinase 2 and 3 (JAK2/JAK3) is one of a number of related tyrosine kinases involved in cytoplasmic signal transduction. In response to a varitety of cytokine or related factors (e.g., interferon, interleukins), JAKs are activated via phosphorylation at 2 adjacent tyrosine residues. The activation of JAKs can lead to the phosphorylation of STAT (signal transducers and activatiors of transcription) proteins, which dimerize and translocate to the nucleus. Once translocated to the nucleus, the STAT proteins can modify transcription of numerous genes, including interferon-stimulated genes. JAK2 is required for the IFN gamma-receptor complex initiation and JAK1 functions as an amplifier. However, active JAK1 may be required for complex responses. Some studies have suggested that the role of JAK2 might be performed by Tyk2 and JAK3, if they were positioned correctly within the IFN gamma-receptor complex.Spezifikation
Phospho-JAK2/JAK3 (Tyr966, Tyr939) | |
Polyclonal | |
Unconjugated | |
JAK2 | |
Breast-JAK3; fae; Fd17; JAK2; JAK-2; JAK3; JAK-3; JAK3_HUMAN; JAK3B; JAKL; Janus kinase 2; Janus kinase 2 (a protein tyrosine kinase); Janus kinase 3; Janus kinase 3 (a protein tyrosine kinase, leukocyte); Janus kinase 3 protein-tyrosine kinase; Janus kinase 3, protein-tyrosine kinase; JTK10; Leukocyte janus kinase; LJAK; L-JAK; RATJAK3; THCYT3; tyrosine-protein kinase JAK2; Tyrosine-protein kinase JAK3; wil | |
Rabbit | |
affinity chromatography | |
RUO | |
16452, 16453, 24514, 25326, 3717, 3718 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O60674, P52333, Q62120, Q62137, Q62689, Q63272 | |
JAK2, JAK3 | |
Synthesized peptide derived from human JAK2/JAK3 around the phosphorylation site of Tyr966/939. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |