Learn More
Phospho-JAK2 (Tyr1007, Tyr1008) Recombinant Polyclonal Antibody (18HCLC), Invitrogen™
Rabbit Recombinant Polyclonal Antibody
Marke: Invitrogen 710928
Additional Details : Gewicht : 0.01000kg
Beschreibung
This antibody is predicted to react with Monkey, Pig, Sheep and Bovine. Recombinant rabbit polyclonal antibodies are unique offerings from Thermo Fisher Scientific. They are comprised of a selection of multiple different recombinant monoclonal antibodies, providing the best of both worlds - the sensitivity of polyclonal antibodies with the specificity of monoclonal antibodies - all delivered with the consistency only found in a recombinant antibody. While functionally the same as a polyclonal antibody - recognizing multiple epitope sites on the target and producing higher detection sensitivity for low abundance targets - a recombinant rabbit polyclonal antibody has a known mixture of light and heavy chains. The exact population can be produced in every lot, circumventing the biological variability typically associated with polyclonal antibody production.
Phospho-JAK2 (Tyr1007, Tyr1008) Recombinant Polyclonal antibody specifically detects Phospho-JAK2 (Tyr1007, Tyr1008) in Human samples. It is validated for Immunocytochemistry, Western Blot.Spezifikation
Phospho-JAK2 (Tyr1007, Tyr1008) | |
Recombinant Polyclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
O60674 | |
JAK2 | |
Peptide corresponding to Human JAK2 (aa 1004-1012) | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Immunocytochemistry, Western Blot | |
18HCLC | |
Unconjugated | |
JAK2 | |
EC 2.7.10.2; Fd17; JAK 2; JAK2; JAK-2; Janus kinase 2; Janus kinase 2 (a protein tyrosine kinase); JTK 10; JTK10; kinase Jak2; THCYT3; Tyrosine-protein kinase JAK2 | |
Rabbit | |
Protein A | |
RUO | |
3717 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |