missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phospho-Fyn (Tyr530) Polyclonal Antibody
GREENER_CHOICE

Artikelnummer. 15543971
Change view
Click to view available options
Menge:
100 μL
Unit Size:
100 Mikroliter
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge unitSize
15543971 100 μL 100 Mikroliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15543971 Supplier Invitrogen™ Supplier No. PA536644

om dit product te kopen Registreer vandaag om een webaccount aan te maken

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This antibody detects endogenous protein at a molecular weight of 59 kDa. Purity is >95% by SDS-PAGE.

FYN is a SRC-related kinase expressed in brain, skeletal muscle and adipose tissues. Fyn functions in a large number of pathways and has been shown to play a role in memory. FYN is a member of the protein-tyrosine kinase oncogene family. It encodes a membrane-associated tyrosine kinase that has been implicated in the control of cell growth. The protein associates with the p85 subunit of phosphatidylinositol 3-kinase and interacts with the fyn-binding protein. Alternatively spliced transcript variants encoding distinct isoforms exist.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-Fyn (Tyr530)
Anwendungen Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Klassifikation Polyclonal
Konzentration 1 mg/mL
Konjugat Unconjugated
Zusammensetzung PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gen Fyn
Gen-Zugriffsnummer P06241, P39688, Q62844
Gen-Alias AI448320; AW552119; C syn protooncogene; c-syn protooncogene; EC 2.7.10.2; Fyn; fyn {ECO:0000312; FYN oncogene related to SRC, FGR, YES; Fyn proto-oncogene; FYN proto-oncogene, Src family tyrosine kinase; kinase Fyn; MGC45350; OKT3-induced calcium influx regulator; OTTHUMP00000017916; OTTHUMP00000017917; P59 p59-Fyn; p59-Fyn; Proto-oncogene c-Fyn; Protooncogene Syn; proto-oncogene Syn; proto-oncogene tyrosine-protein kinase Fyn; RGD:2641}; RP1-66H14.1; SLK; Src Kinase p59; Src like kinase; src/yes-related novel; src-like kinase; SYN; tyrosine kinase p59fyn(T); tyrosine-protein kinase Fyn
Gensymbole Fyn
Wirtsspezies Rabbit
Immunogen Synthetic phosphopeptide derived from human Fyn around the phosphorylation site of Tyrosine 530.
Reinigungsverfahren Antigen affinity chromatography
Menge 100 μL
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 14360, 25150, 2534
Zielspezies Human, Mouse, Rat
Inhalt und Lagerung Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Produkttyp Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.