missing translation for 'onlineSavingsMsg'
Learn More

Phospho-DNA-PK (Thr2647) Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marke:  Invitrogen PA538475

Additional Details : Gewicht : 0.00900kg

Artikelnummer. 15515401

  • 602.00 CHF / 100 Mikrogramm
Verfügbar ab: 12-06-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

The PRKDC gene encodes the catalytic subunit of a nuclear DNA-dependent serine/threonine protein kinase (DNA-PK). The second component is the autoimmune antigen Ku (MIM 152690), which is encoded by the G22P1 gene on chromosome 22q. On its own, the catalytic subunit of DNA-PK is inactive and relies on the G22P1 component to direct it to the DNA and trigger its kinase activity; PRKDC must be bound to DNA to express its catalytic properties.
Spezifikation

Spezifikation

Phospho-DNA-PK (Thr2647)
Polyclonal
Unconjugated
PRKDC
DNA-PKcs, DNAPK, DNPK1, HYRC, HYRC1, XRCC7, p350, DNA-PK catalytic subunit, DNA-dependent protein kinase catalytic subunit, hyper-radiosensitivity of murine scid mutation, complementing 1, p460
Rabbit
Antigen affinity chromatography
RUO
5591
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
1 mg/mL
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4
P78527
PRKDC
A synthetic phosphopeptide derived from human DNA-PK around the phosphorylation site of threonine2647 (T-L-TP-Q-T)
100 μg
Primary
Human
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote