Learn More
Phenylephrone Hydrochloride, TRC
Artikelnummer. 30005432
Change view
Click to view available options
Menge:
25 mg
50 mg
Packungsgröße:
25 Milligramm
50 Milligramm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
30005432 25 mg 25 Milligramm
30005248 50 mg 50 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30005432 Lieferant Toronto Research Chemicals Lieferanten-Nr. TRCP32067025MG
om dit product te kopen Registreer vandaag om een webaccount aan te maken
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemische Identifikatoren
CAS 94240-17-2
Summenformel C9 H11 N O2 . Cl H
Molekulargewicht (g/mol) 201.65
Synonym 1-(3-Hydroxyphenyl)-2-(methylamino)ethanone Hydrochloride, Phenylephrine Hydrochloride Imp. C (EP) as Hydrochloride, Phenylephrone Hydrochloride, Ethanone, 1-(3-hydroxyphenyl)-2-(methylamino)-, hydrochloride (1:1), Ethanone, 1-(3-hydroxyphenyl)-2-(methylamino)-, hydrochloride (9CI)
IUPAC-Name 1-(3-hydroxyphenyl)-2-(methylamino)ethanone;hydrochloride
SMILES Cl.CNCC(=O)c1cccc(O)c1
Spezifikation
Chemischer Name oder Material Phenylephrone Hydrochloride
Hinweise zur Reinheitsqualität HPLC
Menge 25 mg
InChI-Formel InChI=1S/C9H11NO2.ClH/c1-10-6-9(12)7-3-2-4-8(11)5-7;/h2-5,10-11H,6H2,1H3;1H
Formelmasse 201.056
Reinheit (%) >95%