Learn More
PFTK1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA553293
Additional Details : Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: DDTTFDEICV TKMSTRNCQG MDSVIKPLDT IPEDKKVRVQ RTQSTFDPFE KPANQVKRVH SENNACINFK TSSTGKESPK VRRHSSPSSP TSPKFGKADS YEKLEKLGEG SYATVYKGKS KVNG Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 23%.
PFTK1 Polyclonal antibody specifically detects PFTK1 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Spezifikation
PFTK1 | |
Polyclonal | |
Unconjugated | |
Cdk14 | |
Cdk14; Cell division protein kinase 14; cyclin dependent kinase 14; cyclin-dependent kinase 14; hPFTAIRE1; Kiaa0834; mKIAA0834; PFTAIRE protein kinase 1; PFTAIRE1; Pftk1; Serine/threonine-protein kinase PFTAIRE-1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5218 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.10 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O94921 | |
Cdk14 | |
Recombinant protein corresponding to Human PFTK1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only