missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PFTK1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 21612-1-AP-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
This antibody is specific to PFTK1.
PFTK1, a member of the CDC2/CDKX subfamily of Ser/Thr protein kinases, may play a role in meiosis as well as in neuron differentiation and/or function It is highly expressed in brain, pancreas, kidney, heart, testis and ovary, and also detected at lower levels in other tissues except in spleen and thymus where expression is minimal.Spezifikation
PFTK1 | |
Polyclonal | |
Unconjugated | |
CDK14 | |
CDK14, hPFTAIRE1, KIAA0834, PFTAIRE protein kinase 1, PFTAIRE1, PFTK1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
18647, 362316, 5218 | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O35495, O94921 | |
Cdk14 | |
PFTK1 Fusion Protein Ag16259 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Liquid |