missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ PFN1 (Human) Recombinant Protein
Human PFN1 full-length ORF ( AAH06768, 1 a.a. - 140 a.a.) recombinant protein with GST-tag at N-terminal.
Marke: Abnova™ H00005216-P01.25ug
Additional Details : Gewicht : 0.00010kg
Beschreibung
- Sequence: MAGWNAYIDNLMADGTCQDAAIVGYKDSPSVWAAVPGKTFVNITPAEVGVLVGKDRSSFYVNGLTLGGQKCSVIRDSLLQDGEFSMDLRTKSTGGAPTFNVTVTKTDKTLVLLMGKEGVHGGLINKKCYEMASHLRRSQY
Spezifikation
AAH06768 | |
profilin 1 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |
5216 | |
Wheat germ expression system | |
25 μg | |
PFN1 | |
GST |