missing translation for 'onlineSavingsMsg'
Learn More

PEX5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

133.80 CHF - 381.34 CHF

Spezifikation

Antigen PEX5
Konzentration 0.19 mg/mL
Anwendungen Immunoprecipitation, Immunohistochemistry (Paraffin), Immunocytochemistry, Flow Cytometry, Immunofluorescence
Klassifikation Polyclonal
Konjugat Unconjugated
Mehr Spezifikationen ansehen

Produkte 2
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
16850304
Unterlagen ansehen Aktionsdetails
Proteintech
12545-1-AP-20UL
20 μL
133.80 CHF
20 Mikroliter
Alternative Produkte anzeigen
Sparen Sie bis zu 
»
Verfügbar ab: 17-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
 
16840304
Unterlagen ansehen Aktionsdetails
Proteintech
12545-1-AP-150UL
150 μL
381.34 CHF
150 Mikroliter
Alternative Produkte anzeigen
Sparen Sie bis zu 
»
Verfügbar ab: 17-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
 
Beschreibung

Beschreibung

This antibody recognizes endogenous PEX5, which migrates with an apparent molecular mass of 80 kDa.

The product of this gene binds to the C-terminal PTS1-type tripeptide peroxisomal targeting signal (SKL-type) and plays an essential role in peroxisomal protein import. Peroxins (PEXs) are proteins that are essential for the assembly of functional peroxisomes. The peroxisome biogenesis disorders (PBDs) are a group of genetically heterogeneous autosomal recessive, lethal diseases characterized by multiple defects in peroxisome function. The peroxisomal biogenesis disorders are a heterogeneous group with at least 14 complementation groups and with more than 1 phenotype being observed in cases falling into particular complementation groups. Although the clinical features of PBD patients vary, cells from all PBD patients exhibit a defect in the import of one or more classes of peroxisomal matrix proteins into the organelle. Defects in this gene are a cause of neonatal adrenoleukodystrophy (NALD), a cause of Zellweger syndrome (ZWS) as well as may be a cause of infantile Refsum disease (IRD). Alternatively spliced transcript variants encoding different isoforms have been identified.
Spezifikation

Spezifikation

PEX5
Immunoprecipitation, Immunohistochemistry (Paraffin), Immunocytochemistry, Flow Cytometry, Immunofluorescence
Unconjugated
Rabbit
Mouse, Rat, Human
O09012, P50542, Q2M2R8
19305, 312703, 5830
PEX5 Fusion Protein Ag3268
Primary
-20°C
PEX5
0.19 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Peroxin 5, Peroxisome receptor 1, PEX5, PTS1 BP, PTS1 receptor, PTS1R, PXR1
Pex5
IgG
Antigen Affinity Chromatography
Antibody
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Product Certifications