Learn More
DiversifiedBiotech PetriStickers™
GREENER_CHOICE
Artikelnummer. p-200132132
Click to view available options
Enthält:
100 Square Grid
12 Sector Pie
16 Sector Pie
32 Square Grid
50 Square Grid
6 Sector Pie
70 Square Grid
8 Sector Pie
Packungsgröße:
36 Stück
Artikelnummer. 30282323
Marke: DiversifiedBiotech PSTK1000
Nur noch null auf Lager
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Allow for petri dishes to be labeled quickly, neatly and reliably
  • PetriStickers™ allow for petri dishes to be labeled quickly, neatly and reliably
  • Made with water-resistant ink, substrate and adhesives and will remain firmly attached at temperatures from 42° to 4°C
  • Simply remove the label's protective backing and apply it to the back of a petridish
  • After labeling, the dish can be held in a comfortable position for streaking or colony transfer, stored in an incubator or refrigerator, and photographed
Spezifikation
Typ Petri Dish Label
Enthält 50 Square Grid
Durchmesser (metrisch) 7.62 cm