missing translation for 'onlineSavingsMsg'
Learn More

Perfluortetradecansäure, TRC

Artikelnummer. 30007906
Ansicht ändern
Click to view available options
Menge:
500 mg
1 g
5 g
Unit Size:
1 Gramm
5 Gramm
500 Milligramm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge unitSize
30007906 5 g 5 Gramm
30006874 1 g 1 Gramm
30010211 500 mg 500 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 30007906 Supplier Toronto Research Chemicals Supplier No. TRCP2866005G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Hochreine organische Moleküle und analytische Standards, strategisch weltweit bereitgestellt, um Innovation und kommerziellen Erfolg zu ermöglichen.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 376-06-7
Summenformel C14 H F27 O2
Molekulargewicht (g/mol) 714.11
Synonym Tetradecanoic acid, heptacosafluoro- (9CI), 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,14,14,14-Heptacosafluorotetradecanoic acid, PFTeDA, Perfluoro-n-tetradecanoic acid, Perfluoromyristic acid, Perfluorotetradecanecarboxylic acid, Perfluorotetradecanoic acid
IUPAC-Name 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,14,14,14-Heptacosafluortetradecansäure
SMILES OC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

Specifications

Chemischer Name oder Material Perfluorotetradecanoic acid
Hinweise zur Reinheitsqualität HPLC
Menge 5 g
InChI-Formel InChI=1S/C14HF27O2/c15-2(16,1(42)43)3(17,18)4(19,20)5(21,22)6(23,24)7(25,26)8(27,28)9(29,30)10(31,32)11(33,34)12(35,36)13(37,38)14(39,40)41/h(H,42,43)
Formelmasse 713.9545
Reinheit (%) >95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.