missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ PeproGMP® Human Flt-3 Ligand (FLT3L) Recombinant Protein, PeproTech®

Artikelnummer. 30276216
Click to view available options
Menge:
1 mg
100 μg
50 μg
Packungsgröße:
1 Milligramm
100 Mikrogramm
50 Mikrogramm
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Artikelnummer. 30276216

Marke: Gibco™ GMP30019100UG

om dit product te kopen Registreer vandaag om een webaccount aan te maken

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Recombinant Protein

Multiple isoforms of Flt3-Ligand have been identified. The predominant biologically active form is anchored to the cell surface as the extracellular domain of a transmembrane protein (209 a.a.). The membrane-bound isoform can be proteolytically cleaved to generate a biologically active soluble isoform. Additional product information and specifications are provided on the Certificate of Analysis. All PeproGMP® products are manufactured using animal-derived component free materials. Product Description/Intended Use: PeproTech GMP recombinant proteins are manufactured for use as ancillary materials by applying applicable principles of GMP and quality control requirements from USP (United States Pharmacopeia) Chapter 1043 Ancillary Materials for Cell, Gene, and Tissue-Engineered Products. Product use statement: For research use or further manufacturing. Not for diagnostic use or direct administration into humans or animals.

FLT3LG (Fms Related Tyrosine Kinase 3 Ligand) is a Protein Coding gene. Diseases associated with FLT3LG include Aplastic Anemia and Brain Cancer. Among its related pathways are Hematopoietic Stem Cell Differentiation Pathways and Lineage-specific Markers and Ras signaling pathway. Gene Ontology (GO) annotations related to this gene include protein homodimerization activity and cytokine activity. Dendritic cells (DCs) provide the key link between innate and adaptive immunity by recognizing pathogens and priming pathogen-specific immune responses. FLT3LG controls the development of DCs and is particularly important for plasmacytoid DCs and CD8-positive classical DCs and their CD103-positive tissue counterparts.
TRUSTED_SUSTAINABILITY

Spezifikation

Zugriffsnummer P49771
Zur Verwendung mit (Anwendung) Control, Western Blot Control, Bioactivity
Format Lyophilized
Zusammensetzung Protein with no preservative
Gen-ID (Entrez) 2323
Molekulargewicht 17.6 kDa
Name PeproGMP™ Human Flt-3 Ligand (FLT3L)
Reinigungsverfahren Purified
Menge 100 μg
Lagerungsbedingungen -20°C or -80°C if preferred
Kennzeichnung GMP
Endotoxin-Konzentration ≤0.1 EU/μg
Gen-Alias FL; Flt 3 ligand; Flt3; Flt3 L; Flt3 ligand; Flt3l; Flt3lg; fms related receptor tyrosine kinase 3 ligand; fms related tyrosine kinase 3 ligand; FMS-like tyrosine kinase 3 ligand; fms-related tyrosine kinase 3; fms-related tyrosine kinase 3 ligand; H-FLT-3-L; Ly72L; M-FLT-3-L; NEWGENE_2322792; SL cytokine
Gebräuchliche Bezeichnung FLT3LG
Gensymbol FLT3LG
Biologische Aktivität Determined by its ability to stimulate human AML5 cell proliferation. PeproGMP Recombinant Human Flt3-Ligand has been tested against the WHO standard NIBSC 96/532.
Produkttyp Protein
Konjugat Unconjugated
Rekombinant Recombinant
Sequenz MTQDCSFQHS PISSDFAVKI RELSDYLLQD YPVTVASNLQ DEELCGGLWR LVLAQRWMER LKTVAGSKMQ GLLERVNTEI HFVTKCAFQP PPSCLRFVQT NISRLLQETS EQLVALKPWI TRQNFSRCLE LQCQPDSSTL PPPWSPRPLE ATAPTA
Inhalt und Lagerung -20°Cor to +80°C if preferred
Aktivität Determined by its ability to stimulate human AML5 cell proliferation. PeproGMP Recombinant Human Flt3-Ligand has been tested against the WHO standard NIBSC 96/532. Find the International Unit (IU) value <a href='https://www.thermofisher.com/us/en/home/life-science/cell-culture/cell-culture-learning-center/recombinant-protein-information/international-units.html'>here</a>.
Expressionssystem E. coli
Form Lyophilized
Produktlinie PeproGMP
Reinheits- oder Qualitätsgrad ≥ 98% by SDS-PAGE gel and HPLC analyses.
Mehr anzeigen Weniger anzeigen
Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt