missing translation for 'onlineSavingsMsg'
Learn More
Gibco™ PeproGMP® Human FGF-basic (FGF-2/bFGF) Recombinant Protein, PeproTech® Artikelnummer.: 30276681
Nur noch undefined auf Lager
Zum Warenkorb hinzufügen

Gibco™ PeproGMP® Human FGF-basic (FGF-2/bFGF) Recombinant Protein, PeproTech®

Artikelnummer. p-200088553
1 mg, 1 Milligramm
Click to view available options
Menge:
1 mg
100 μg
Packungsgröße:
1 Milligramm
100 Mikrogramm
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Artikelnummer. 30276681

Marke: Gibco™ GMP10018B1MG

Nur noch undefined auf Lager
Zum Warenkorb hinzufügen
Nur noch undefined auf Lager
Zum Warenkorb hinzufügen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Recombinant Protein

Additional product information and specifications are provided on the Certificate of Analysis. All PeproGMP® products are manufactured using animal-derived component free materials. Product Description/Intended Use: PeproTech GMP recombinant proteins are manufactured for use as ancillary materials by applying applicable principles of GMP and quality control requirements from USP (United States Pharmacopeia) Chapter 1043 Ancillary Materials for Cell, Gene, and Tissue-Engineered Products. Product use statement: For research use or further manufacturing. Not for diagnostic use or direct administration into humans or animals.

FGF2 (FGFb, fibroblast growth factor basic) belongs to the fibroblast growth factor (FGF) family, and interacts with high-affinity transmembrane receptors to influence cell proliferation and tissue neovascularization. FGF2 exists as five isoforms with distinct intracellular localizations and functions. The 18 kDa isoform is predominantly cytosolic and acts through cell surface receptors, whereas the 22, 22.5, 24 and 34 kDa isoforms are nuclear and may signal independent of transmembrane receptor pathways. In humans, the gene is located on the q arm of chromosome 4. FGF2 has been implicated in diverse biological processes, such as limb and nervous system development, wound healing, and tumor growth. The mRNA for FGF2 contains multiple polyadenylation sites, and is alternatively translated from non-AUG and AUG initiation codons, resulting in five different isoforms with distinct properties. The CUG-initiated isoforms are localized in the nucleus and are responsible for the intracrine effect, whereas, the AUG-initiated form is mostly cytosolic and is responsible for the paracrine and autocrine effects of this FGF. Diseases associated with FGF2 dysfunction include Kaposi Sarcoma and corneal neovascularization.
TRUSTED_SUSTAINABILITY

Spezifikation

Zugriffsnummer P09038
Zur Verwendung mit (Anwendung) Functional Assay
Format Lyophilized
Zusammensetzung protein with no preservative
Gen-ID (Entrez) 2247
Molekulargewicht 17.2 kDa
Name PeproGMP™ Human FGF-basic (FGF-2/bFGF)
Menge 1 mg
Kennzeichnung RUO
Endotoxin-Konzentration <0.1 EU/ μg
Gen-Alias Basic fibroblast growth factor; basic fibroblast growth factor bFGF; bFGF; FGF; fgf basic; Fgf2; Fgf-2; Fgfb; FGF-b; Fibroblast growth factor; fibroblast growth factor 2; fibroblast growth factor 2 (basic); HBGF-2; Heparin-binding growth factor 2; H-FGF-b-147; H-FGF-b-154; M-FGF-b; Prostatic growth factor; prostatropin
Gebräuchliche Bezeichnung FGF2
Biologische Aktivität Determined by the proliferation of mouse Balb/c 3T3 cells. PeproGMP Recombinant Human FGF-basic has been tested against the WHO standard NIBSC 90/712.
Konjugat Unconjugated
Rekombinant Recombinant
Sequenz AAGSITTLPA LPEDGGSGAF PPGHFKDPKR LYCKNGGFFL RIHPDGRVDG VREKSDPHIK LQLQAEERGV VSIKGVCANR YLAMKEDGRL LASKCVTDEC FFFERLESNN YNTYRSRKYT SWYVALKRTG QYKLGSKTGP GQKAILFLPM SAKS
Inhalt und Lagerung -20°Cor to +80°C if preferred
Aktivität Determined by the proliferation of mouse Balb/c 3T3 cells. PeproGMP Recombinant Human FGF-basic has been tested against the WHO standard NIBSC 90/172.
Expressionssystem E. coli
Form Lyophilized
Produktlinie PeproGMP
Reinheits- oder Qualitätsgrad ≥ 95% by SDS-PAGE gel and HPLC analyses.
Mehr anzeigen Weniger anzeigen

Zertifikate

Eine Chargennummer ist erforderlich, um Ergebnisse für Zertifikate anzuzeigen. Um Ihre Chargennummer zu früheren Bestellungen zu finden, können Sie unsere Auftragsverfolgung nutzen.

5 Ergebnisse gefunden
Chargennummer Art des Zertifikats Datum Artikelnummer
Chargennummer1024G008-1J1624 Art des ZertifikatsAnalysenzertifikat Datum16/07/2025 ArtikelnummerGMP100-18B-1MG
Chargennummer1024G008-1J1424 Art des ZertifikatsAnalysenzertifikat Datum16/07/2025 ArtikelnummerGMP100-18B-1MG
Chargennummer1223G008A3124 Art des ZertifikatsAnalysenzertifikat Datum16/07/2025 ArtikelnummerGMP100-18B-1MG
Chargennummer1223G008A2224 Art des ZertifikatsAnalysenzertifikat Datum16/07/2025 ArtikelnummerGMP100-18B-1MG
Chargennummer1124G008L1624 Art des ZertifikatsAnalysenzertifikat Datum16/07/2025 ArtikelnummerGMP100-18B-1MG
Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts
Gibco™ PeproGMP® Human FGF-basic (FGF-2/bFGF) Recombinant Protein, PeproTech® >

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt