missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ PeproGMP™ Human BMP-4 Recombinant Protein, PeproTech®

Artikelnummer. 30275254
Click to view available options
Menge:
100 μg
50 μg
Packungsgröße:
100 Mikrogramm
50 Mikrogramm
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Artikelnummer. 30275254

Marke: Gibco™ GMP12005ET100UG

om dit product te kopen Registreer vandaag om een webaccount aan te maken

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Recombinant Protein

Bone morphogenetic proteins (BMPs) constitute a subfamily within the TGF-beta superfamily of structurally related signaling proteins. PeproTech's E. coli-derived Human BMP-4 is a fully active homodimeric protein consisting of two 107 amino acid subunits, which correspond to amino acids 303-408 of the full-length BMP-4 precursor, as well as an N-terminal methionine. PeproTech Recombinant Human BMP-4 is a 24.2 kDa protein. PeproGMP BMP-4 can be used with PeproGMP VEGF-165 and PeproGMP SCF in CTS StemScale medium for pluripotent stem cell differentiation, specifically for mesoderm and hematopoetic progenitor cell induction. Reconstitute using 5mM HCl, pH 3.0 to a concentration of 0.1-1.0 mg/ml. Additional product information, specifications, handling and reconstitution information are provided on the Certificate of Analysis. All PeproGMP™ products are manufactured using animal-derived component free materials. Product Description/Intended Use: PeproTech GMP recombinant proteins are manufactured for use as ancillary materials by applying applicable principles of GMP and quality control requirements from USP (United States Pharmacopeia) Chapter <1043> Ancillary Materials for Cell, Gene, and Tissue-Engineered Products.

The protein encoded by this gene is a member of the bone morphogenetic protein family which is part of the transforming growth factor-beta superfamily. The superfamily includes large families of growth and differentiation factors. Bone morphogenetic proteins were originally identified by an ability of demineralized bone extract to induce endochondral osteogenesis in vivo in an extraskeletal site. This particular family member plays an important role in the onset of endochondral bone formation in humans, and a reduction in expression has been associated with a variety of bone diseases, including the heritable disorder Fibrodysplasia Ossificans Progressiva. Alternative splicing in the 5' untranslated region of this gene has been described and three variants are described, all encoding an identical protein.
TRUSTED_SUSTAINABILITY

Spezifikation

Zugriffsnummer P12644
Zur Verwendung mit (Anwendung) Control, Western Blot Control, Bioactivity
Format Lyophilized
Zusammensetzung Protein with no preservative
Gen-ID (Entrez) 652
Molekulargewicht 24.2 kDa
Name PeproGMP™ Human BMP-4
Reinigungsverfahren Purified
Menge 100 μg
Lagerungsbedingungen -20°C or -80°C if preferred
Kennzeichnung GMP
Endotoxin-Konzentration ≤0.1 EU/μg
Gen-Alias BMP; BMP2B; BMP-2B; Bmp2b1; Bmp2b-1; Bmp4; Bmp-4; BOMPR4A; Bone morphogenetic protein; Bone morphogenetic protein 2B; bone morphogenetic protein 4; bone morphogenetic protein 4 preproprotein; DVR4; Dvr-4; MCOPS6; OFC11; ZYME
Gebräuchliche Bezeichnung BMP4
Gensymbol BMP4
Biologische Aktivität Determined by its ability to induce alkaline phosphatase production by ATDC-5 cells.
Produkttyp Protein
Konjugat Unconjugated
Rekombinant Recombinant
Sequenz MKKNKNCRRH SLYVDFSDVG WNDWIVAPPG YQAFYCHGDC PFPLADHLNS TNHAIVQTLV NSVNSSIPKA CCVPTELSAI SMLYLDEYDK VVLKNYQEMV VEGCGCR
Inhalt und Lagerung -20°Cor to +80°C if preferred
Aktivität Determined by its ability to induce alkaline phosphatase production by ATDC-5 cells.
Expressionssystem E. coli
Form Lyophilized
Produktlinie PeproGMP
Reinheits- oder Qualitätsgrad ≥ 98% by SDS-PAGE gel and HPLC analyses.
Mehr anzeigen Weniger anzeigen
Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt