missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pellino 1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
249.00 CHF - 573.00 CHF
Spezifikation
| Antigen | Pellino 1 |
|---|---|
| Verdünnung | Western Blot 1:500-1:1000, Immunohistochemistry-Paraffin 1:100 |
| Anwendungen | Western Blot, Immunohistochemistry (Paraffin) |
| Klassifikation | Polyclonal |
| Konjugat | Unconjugated |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|
18065665
|
Novus Biologicals
NB100-56672SS |
0.05 mL |
249.00 CHF
0.05 Milliliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
|
18730174
|
Novus Biologicals
NB100-56672 |
573.00 CHF
0.20 Milliliter |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | ||||||
Beschreibung
Pellino 1 Polyclonal specifically detects Pellino 1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Spezifikation
| Pellino 1 | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| DKFZp686C18116, MGC50990, pellino (Drosophila) homolog 1, pellino homolog 1 (Drosophila), pellino-1, Pellino-related intracellular-signaling molecule, PRISM, protein pellino homolog 1 | |
| PELI1 | |
| IgG | |
| Unpurified |
| Western Blot 1:500-1:1000, Immunohistochemistry-Paraffin 1:100 | |
| Polyclonal | |
| Antisera | |
| RUO | |
| Whole antisera with PBS and 0.05% BSA with 0.05% Sodium Azide | |
| 57162 | |
| This antibody was developed against KLH-conjugated synthetic peptide corresponding to a portion of amino acids 100-150 of human Pellino 1 (Peli1), Genbank No. NP_065702.2. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts