missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDZD2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 20039-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
The antibody recognizes a region near the N-term of PDZD2.
Proteins containing PDZ domains have been shown frequently to bind the C-termini of transmembrane receptors or ion channels. They have also been shown to bind to other PDZ domain proteins and could possibly be involved in intracellular signaling. The protein encoded by this gene contains six PDZ domains and shares sequence similarity with pro-interleukin-16 (pro-IL-16). Like pro-IL-16, the encoded protein localizes to the endoplasmic reticulum and is thought to be cleaved by a caspase to produce a secreted peptide containing two PDZ domains. In addition, this gene is upregulated in primary prostate tumors and may be involved in the early stages of prostate tumorigenesis.Spezifikation
PDZD2 | |
Polyclonal | |
Unconjugated | |
PDZD2 | |
AIPC, KIAA0300, PAPIN, PDZ domain containing 2, PDZD2, PDZK3, PIN1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23037, 65034, 68070 | |
Antibody | |
IgG |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O15018, Q9QZR8 | |
Pdzd2 | |
Peptide | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Liquid |