missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDLIM1/CLP36 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 11674-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
Initiation of transcription by RNA polymerase II requires the activities of more than 70 polypeptides. The protein that coordinates these activities is transcription factor IID (TFIID), which binds to the core promoter to position the polymerase properly, serves as the scaffold for assembly of the remainder of the transcription complex, and acts as a channel for regulatory signals. TFIID is composed of the TATA-binding protein (TBP) and a group of evolutionarily conserved proteins known as TBP-associated factors or TAFs. TAFs may participate in basal transcription, serve as coactivators, function in promoter recognition or modify general transcription factors (GTFs) to facilitate complex assembly and transcription initiation. A distinctive feature of TBP is a long string of glutamines in the N-terminal. This region of the protein modulates the DNA binding activity of the C terminus, and modulation of DNA binding affects the rate of transcription complex formation and initiation of transcription. Mutations that expand the number of CAG repeats encoding this polyglutamine tract, and thus increase the length of the polyglutamine string, are associated with spinocerebellar ataxia 17, a neurodegenerative disorder classified as a polyglutamine disease.Spezifikation
PDLIM1/CLP36 | |
Polyclonal | |
Unconjugated | |
PDLIM1 | |
CLIM1, CLP 36, CLP36, Elfin, hCLIM1, LIM domain protein CLP 36, PDLIM1, PDZ and LIM domain 1, PDZ and LIM domain protein 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
54132, 54133, 9124 | |
-20°C | |
Liquid |
Immunofluorescence, Immunoprecipitation, Flow Cytometry, Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.6 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O00151, O70400, P52944 | |
Pdlim1 | |
PDLIM1/CLP36 Fusion Protein Ag2274 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |