missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDK4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 12949-1-AP-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
PDK4 is a mitochondrial protein with a histidine kinase domain that is a member of the PDK/BCKDK protein kinase family. This protein is located in the matrix of the mitrochondria and it′s expression is regulated by glucocorticoids, retinoic acid and insulin. PDK4 inhibits the mitochondrial pyruvate dehydrogenase complex by phosphorylation of the E1 alpha subunit, thus contributing to the regulation of glucose metabolism.Spezifikation
PDK4 | |
Polyclonal | |
Unconjugated | |
PDK4 | |
PDK4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
27273, 5166, 89813 | |
Antibody | |
IgG |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O54937, O70571, Q16654 | |
Pdk4 | |
PDK4 Fusion Protein Ag3629 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Liquid |