missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDK3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 12215-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
PDK3 inhibits the mitochondrial pyruvate dehydrogenase complex by phosphorylation of the E1 alpha subunit, thus contributing to the regulation of glucose metabolism.Spezifikation
PDK3 | |
Polyclonal | |
Unconjugated | |
PDK3 | |
PDK3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
236900, 5165 | |
Antibody | |
IgG |
Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation | |
0.35 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q15120, Q922H2 | |
PDK3 | |
PDK3 Fusion Protein Ag2879 | |
20 μL | |
Primary | |
Human, Mouse | |
Liquid |