missing translation for 'onlineSavingsMsg'
Learn More

PDIA6 Mouse anti-Human, Mouse, Rat, Clone: 1G2D4, Proteintech

Mouse Monoclonal Antibody

133.80 CHF - 381.34 CHF

Spezifikation

Antigen PDIA6
Klon 1G2D4
Konzentration 1 mg/mL
Anwendungen Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry, Western Blot
Klassifikation Monoclonal
Mehr Spezifikationen ansehen

Produkte 2
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
16848765
Unterlagen ansehen Aktionsdetails
Proteintech
66669-1-IG-20UL
20 μL
133.80 CHF
20 Mikroliter
Alternative Produkte anzeigen
Sparen Sie bis zu 
»
Verfügbar ab: 16-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
 
16838765
Unterlagen ansehen Aktionsdetails
Proteintech
66669-1-IG-150UL
150 μL
381.34 CHF
150 Mikroliter
Alternative Produkte anzeigen
Sparen Sie bis zu 
»
Verfügbar ab: 16-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
 
Description

Description

This antibody is specific to PDIA6.

P5 is a novel protein that was originally identified in humans. Functionally, P5 has been shown to have peptide binding ability and chaperone activity specific to certain proteins. P5 shares several structural similarities to PDI, an important protein of the ER. The deduced amino acid sequence of P5 shows that it contains the ER retention signal KDEL. P5 also contains two CXXC-like motifs, a motif responsible for oxidoreductase activity. These two CXXC motifs are found on the N- and C-terminus of the protein, one at each end. Studies suggest that the N-terminal CXXC motif is more important than the second for isomerase activity. Contrary to most of the PDI-like proteins, P5 is not stress-inducible.
Spécification

Spécification

PDIA6
1 mg/mL
Monoclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
ER protein 5, ERP5, P5, PDIA6, Protein disulfide isomerase A6, Protein disulfide isomerase P5, TXNDC7
Pdia6
IgG1
Protein G
Antibody
1G2D4
Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry, Western Blot
Unconjugated
Mouse
Rat, Human, Mouse
Q15084, Q63081, Q922R8
10130, 286906, 71853
PDIA6 Fusion Protein Ag27608
Primary
-20°C
PDIA6
Vidéos
FDS
Documents

Documents

Product Certifications