missing translation for 'onlineSavingsMsg'
Learn More

PDGFRβ Ab-1, Rabbit Polyclonal Antibody,Lab Vision™ Epredia™

Provide accurate, reproducible results with the Epredia™ PDGFRβ Ab-1, Rabbit Polyclonal Antibody.

auf Anfrage

Spezifikation

Konzentration 1mg/mL
Anwendungen Immunohistochemistry (Paraffin)
Klassifikation Polyclonal
Konjugat Unconjugated
Form Concentrated
Mehr Spezifikationen ansehen

Beschreibung

Beschreibung

PDGFR (Platelet Derived Growth Factor Receptor) exhibits two different isoforms, α and β coded by two different genes. They possess five immunoglobulin like domains that are involved in ligand binding. Two tyrosine kinase domains are separated by a kinase insert to which PI-3 kinase can bind. PDGF AA can bind autophosphorylate and activate only PDGFR, α while PDGF BB can bind and activate to both PDGFR, α and β.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human and Mouse. Others not known.

Epitope: Near C-terminal

Immunogen: A synthetic peptide derived from near the C-terminus of human PDGFRβ

Molecular Weight: ∽190kDa

Postitive Control: Ovarian CA

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO

Spezifikation

Spezifikation

1mg/mL
Polyclonal
Concentrated
RUO
Human
IgG
Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Growth Factor
A synthetic peptide derived from near the C-terminus of human PDGFR, beta
Primary
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Product Certifications