Learn More
Cole Parmer PCRmax Alpha 1 Thermal Cycler, Single 96-Well Capacity; 100 to 230 VAC
Marke:  Cole Parmer 93945-02
PCRmax Alpha 1 Thermal Cycler, Single 96-Well Capacity; 100 to 230 VAC
Artikelnummer. 15601979
  • ONLINE EXCLUSIVE PRICE
    8250.00 CHF
    5895.63 CHF / 1 Stück
    Sparen 2354.37 CHF
    28% Rabatt
Verfügbar ab: 23-12-2026