missing translation for 'onlineSavingsMsg'
Learn More

PASD1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marke:  Proteintech 12976-1-AP-150UL

Additional Details : Gewicht : 0.00010kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 16840964

  • 351.68 CHF / 150 Mikroliter

Verfügbar ab: 28-06-2022
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

This gene encodes a protein that is thought to function as a transcription factor. The protein is a cancer-associated antigen that can stimulate autologous T-cell responses, and it is therefore considered to be a potential immunotherapeutic target for the treatment of various hematopoietic maligcies, including diffuse large B-cell lymphoma.
Spezifikation

Spezifikation

PASD1
Polyclonal
Unconjugated
PASD1
Cancer/testis antigen 63, CT63, OX TES 1, PAS domain containing 1, PASD1
Rabbit
Antigen Affinity Chromatography
RUO
139135, 382221
Antibody
IgG
Western Blot
0.4 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8IV76
Gm1141, PASD1
PASD1 Fusion Protein Ag3587
150 μL
Primary
Human, Mouse
Liquid
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Zertifikate
Sonderangebote

Sonderangebote