missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PARK2/Parkin Mouse anti-Human, Mouse, Clone: 2H5A7, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech 66674-1-IG-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
The precise function of Parkin gene is unknown; however, the encoded protein is a component of a multiprotein E3 ubiquitin ligase complex that mediates the targeting of substrate proteins for proteasomal degradation. Mutations in this gene are known to cause Parkinson disease and autosomal recessive juvenile Parkinson disease. Alternative splicing of this gene produces multiple transcript variants encoding distinct isoforms. Additional splice variants of this gene have been described but currently lack transcript support.Spezifikation
PARK2/Parkin | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O60260 | |
Park2, PRKN | |
PARK2/Parkin Fusion Protein Ag5179 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG2b |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
2H5A7 | |
Unconjugated | |
PRKN | |
AR JP, LPRS2, PARK2, Parkinson disease protein 2, PDJ, PRKN | |
Mouse | |
Protein A | |
RUO | |
5071, 50873 | |
-20°C | |
Liquid |