missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAR2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 12160-1-AP-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
Coagulation factor II (thrombin) receptor-like 1 (F2RL1) is a member of the large family of 7-transmembrane-region receptors that couple to guanosine-nucleotide-binding proteins. F2RL1 is also a member of the protease-activated receptor family. It is activated by trypsin, but not by thrombin. It is activated by proteolytic cleavage of its extracellular amino terminus. The new amino terminus functions as a tethered ligand and activates the receptor. The F2RL1 gene contains two exons and is widely expressed in human tissues. The predicted protein sequence is 83% identical to the mouse receptor sequence.Spezifikation
PAR2 | |
Polyclonal | |
Unconjugated | |
F2RL1 | |
F2RL1, G protein coupled receptor 11, GPR11, PAR 2, PAR2, Thrombin receptor like 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
116677, 14063, 2150 | |
-20°C | |
Liquid |
Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P55085, P55086, Q63645 | |
F2RL1 | |
PAR2 Fusion Protein Ag2801 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |