missing translation for 'onlineSavingsMsg'
Learn More

PAR2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marke:  Proteintech 12160-1-AP-20UL

Additional Details : Gewicht : 0.00010kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 16839703

  • 133.80 CHF / 20 Mikroliter
Verfügbar ab: 17-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Coagulation factor II (thrombin) receptor-like 1 (F2RL1) is a member of the large family of 7-transmembrane-region receptors that couple to guanosine-nucleotide-binding proteins. F2RL1 is also a member of the protease-activated receptor family. It is activated by trypsin, but not by thrombin. It is activated by proteolytic cleavage of its extracellular amino terminus. The new amino terminus functions as a tethered ligand and activates the receptor. The F2RL1 gene contains two exons and is widely expressed in human tissues. The predicted protein sequence is 83% identical to the mouse receptor sequence.
Spezifikation

Spezifikation

PAR2
Polyclonal
Unconjugated
F2RL1
F2RL1, G protein coupled receptor 11, GPR11, PAR 2, PAR2, Thrombin receptor like 1
Rabbit
Antigen Affinity Chromatography
RUO
116677, 14063, 2150
-20°C
Liquid
Western Blot
0.23 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P55085, P55086, Q63645
F2RL1
PAR2 Fusion Protein Ag2801
20 μL
Primary
Mouse, Rat, Human
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote