missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAK2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 19979-1-AP-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
The antibody has no cross reaction to PAK1 and PAK3.
NF2 encodes a protein, Merlin, that is similar to some members of the ERM family of proteins that are thought to link cytoskeletal components with proteins in the cell membrane. Merlin has been shown to interact with cell-surface proteins, proteins involved in cytoskeletal dynamics and proteins involved in regulating ion transport. This gene is expressed at high levels during embryonic development; in adults, significant expression is found in Schwann cells, meningeal cells, lens and nerve. Mutations in this gene are associated with neurofibromatosis type II which is characterized by nervous system and skin tumors and ocular abnormalities. Two predomit isoforms and a number of minor isoforms are produced by alternatively spliced transcripts.Spezifikation
PAK2 | |
Polyclonal | |
Unconjugated | |
PAK2 | |
C t PAK2, Gamma PAK, p21 activated kinase 2, p27 PAK 2p34, p34, p58, PAK 2, PAK2, PAK65, PAKgamma, S6/H4 kinase | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
5062 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q13177 | |
PAK2 | |
Peptide | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |